missing translation for 'onlineSavingsMsg'
Learn More

Decyl sodium sulfate, HPLC grade

Product Code. 10400841 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10400841 5 g Glass bottle 5g
10420841 25 g Glass bottle 25g
10723734 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10400841 Supplier Thermo Scientific Supplier No. 406220050
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142-87-0 | C10H19Na·H2SO4 | 260.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-87-0
MDL Number MFCD00041881
InChI Key XZTJQQLJJCXOLP-UHFFFAOYSA-M
Synonym sodium decyl sulfate, sodium n-decyl sulfate, decyl sodium sulfate, sulfuric acid, monodecyl ester, sodium salt, sodium decyl sulphate, unii-al92m833sy, n-decyl sodium sulfate, sodium n-decylsulphate, sulfuric acid, monodecyl ester, sodium salt 1:1, sulfuric acid, decyl ester, sodium salt
PubChem CID 23665771
IUPAC Name sodium;decyl sulfate
SMILES CCCCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Absorbance (0.005M in water) (1 cm cell), (0.005 M in water) (1 cm cell) at 0nm, 0.01 max. at 254nm, 0.01 max. at 280nm, 0.05 max. at 220nm, 0.1 max. at 200nm
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C10H21NaO4S
Linear Formula CH3(CH2)9OSO3Na
Quantity 5 g
Molecular Weight (g/mol) 260.324
Formula Weight 260.32
Percent Purity ≥98.5% (titrimetric) (on dry substance)
Grade HPLC
Physical Form Powder
Chemical Name or Material Decyl sodium sulfate
Show More Show Less
compliance-icons
Product Identifier
  • Decyl sodium sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.