missing translation for 'onlineSavingsMsg'
Learn More

Decyl beta-D-maltopyranoside, 97+%

Product Code. 15418089
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
This item is not returnable. View return policy

Product Code. 15418089

Brand: Thermo Scientific Alfa Aesar J66644.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82494-09-5 | C22H42O11 | 482.567 g/mol

Decyl beta-D-maltopyranoside is used in a study to assess α-helical membrane protein crystallization. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography. It aids in solubilizing the active cytochrome oxidase from mitochondria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decyl beta-D-maltopyranoside is used in a study to assess α-helical membrane protein crystallization. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography. It aids in solubilizing the active cytochrome oxidase from mitochondria.

Solubility
Soluble in methanol at 50mg/ml

Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82494-09-5
Molecular Formula C22H42O11
Molecular Weight (g/mol) 482.567
MDL Number MFCD00061624
InChI Key WOQQAWHSKSSAGF-WXFJLFHKSA-N
Synonym decyl beta-d-maltopyranoside, decyl 4-o-a-d-glucopyranosyl-b-d-glucopyranoside, n-decyl-b-d-maltoside, decyl-, a-d-maltoside, decyl-beta-d-maltopyranoside, n-decyl beta-d-maltopyranoside, decyl 4-o-alpha-d-glucopyranosyl-beta-d-glucopyranoside, decyl alpha-d-glucopyranosyl-1->4-beta-d-glucopyranoside, n-decyl-beta-d-maltopyranoside, decyl b-d-maltopyranoside
PubChem CID 5288728
ChEBI CHEBI:67097
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-decoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

Color White
Quantity 5 g
Beilstein 4923334
Solubility Information Soluble in methanol at 50mg/ml
Formula Weight 482.56
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Decyl beta-D-maltopyranoside

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.