missing translation for 'onlineSavingsMsg'
Learn More

Decyl beta-D-glucopyranoside, Thermo Scientific Chemicals

Product Code. 15417779
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417779 1 g 1g
15427779 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417779 Supplier Thermo Scientific Alfa Aesar Supplier No. J66423.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58846-77-8
Molecular Formula C16H32O6
Molecular Weight (g/mol) 320.426
MDL Number MFCD00063297
InChI Key JDRSMPFHFNXQRB-IBEHDNSVSA-N
Synonym decyl beta-d-glucopyranoside, n-decyl-b-d-glucopyranoside, decyl-beta-d-glucopyranoside, n-decyl-beta-d-glucopyranoside, unii-z17h97ea6y, n-decyl glucoside, decyl beta-d-glucoside, triton x 190, 28-deacetylsendanin, beta-d-glucoside, decyl
PubChem CID 62142
IUPAC Name (2R,3R,4S,5S,6R)-2-decoxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O

Specifications

Color White
Quantity 1 g
Beilstein 85089
Solubility Information Soluble in methanol
Formula Weight 320.42
Physical Form Powder
Chemical Name or Material Decyl beta-D-glucopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.