missing translation for 'onlineSavingsMsg'
Learn More

Decyl alcohol, 98+%, Thermo Scientific Chemicals

Catalog Number p-3736091
30.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
112-30-1
C10H22O
158.28
MFCD00004747
MWKFXSUHUHTGQN-UHFFFAOYSA-N
1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac
8174
CHEBI:28903
decan-1-ol
CCCCCCCCCCO
112-30-1
C10H22O
158.28
MFCD00004747
MWKFXSUHUHTGQN-UHFFFAOYSA-N
1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac
8174
CHEBI:28903
decan-1-ol
CCCCCCCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 6°C
Color Colorless to Yellow
Density 0.8290g/mL
Boiling Point 220°C to 240°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)9OH
Refractive Index 1.4360 to 1.438
Quantity 1 L
Beilstein 01, 425
Merck Index 15, 2858
Specific Gravity 0.829
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,chlorogorm,soluble in alcohol,ether,mineral oil,propylene,insoluble in glycerin water (233°C)
Viscosity 14.1 mPa.s (20°C)
Formula Weight 158.28
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Decyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Decyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'