missing translation for 'onlineSavingsMsg'
Learn More

Decursin, MedChemExpress

Product Code. 16483626
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483626

Brand: Medchem Express HY18981

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Decursin ((+)-Decursin) is a cytotoxic agent and a potent protein kinase C activator from the Root of Angelica gigas. Decursin inhibits tumor growth, migration, and invasion in gastric cancer by down-regulating CXCR7 expression. Decursin inhibits the growth of HepG2 cells via Hippo/YAP signaling pathway.

Decursin ((+)-Decursin) is a cytotoxic agent and a potent protein kinase C activator from the Root of Angelica gigas. Decursin inhibits tumor growth, migration, and invasion in gastric cancer by down-regulating CXCR7 expression. Decursin inhibits the growth of HepG2 cells via Hippo/YAP signaling pathway.

Specifications

Chemical Name or Material Decursin
CAS 5928-25-6
Color White
Purity Grade Notes Research
Molecular Formula C19H20O5
Quantity 25 mg
Synonym (+)-Decursin
Solubility Information DMSO : 50 mg/mL (152.27 mM; Need ultrasonic)
SMILES C/C(C)=C\C(O[C@H](C(C)(C)O1)CC2=C1C=C(O3)C(C=CC3=O)=C2)=O
Molecular Weight (g/mol) 328.36
Formula Weight 328.36
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.