missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dectin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSD00004W
This item is not returnable.
View return policy
Description
Reconstitute with 100 μL of distilled water.
Specifications
| Dectin 2 | |
| Polyclonal | |
| whole serum with no preservative | |
| Q6EIG7 | |
| CLEC6A | |
| A synthetic peptide from n-termnal region of human Dectin-2 | |
| RUO | |
| 93978 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| CLEC6A | |
| Dendritic cell-associated lectin 2, DC-associated C-type lectin 2, CLEC6A, Dectin 2, C-type lectin superfamily member 10, CLECSF10, DECTIN2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction