missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dectin 2 Monoclonal Antibody (3D1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516250
This item is not returnable.
View return policy
Description
Sequence homology: Bovine/Sheep (83%), Mouse (65%), Rat (52%).
Dectin-2 is a C-type lectin expressed by dendritic cells, macrophages and inflammatory monocytes. It is a type II membrane receptor, with a short cytoplasmic tail, and a C-terminal C-type lectin domain. Dectin-2 is a receptor with specificity for high mannose structures and mediates cytokine production from DC and other antigen presenting cells. Though, Dectin-2 has no obvious signaling motifs attached to CP tail, reports indicate that Dectin-2 employs FcRg for internalization and to activate NF-kappaB pathway for cytokine production. Though it shares only 20-25% of homology with Dectin-1, it functions similarly as pattern recognition receptor (PRR) for fungal infections, to activate both innate (Syk-CARD9 innate signaling pathway) and adaptive immune responses (by activation of DC) to fungal infections. Thus Dectin-2 is a connector molecule of both innate and adaptive immunity functioning at DC level.
Specifications
| Dectin 2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6EIG7 | |
| CLEC6A | |
| A blend of two peptides made to amino acids 44-63 and 90-106 of human Dectin-2. | |
| 100 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG3 κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 3D1 | |
| Unconjugated | |
| CLEC6A | |
| Clec4n; CLEC6A; Clec6a-ps1; CLECSF10; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 10; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 10; C-type lectin domain family 4, member n; C-type lectin domain family 6 member A; C-type lectin domain family 6, member A; C-type lectin domain family 6, member A, pseudogene 1; C-type lectin superfamily member 10; DC-associated C-type lectin 2; dectin 2; DECTIN2; dectin-2; Dectin-2p; Dendritic cell-associated C-type lectin 2; natural killer complex lectin; Nkcl | |
| Mouse | |
| Protein G | |
| RUO | |
| 93978 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction