missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dectin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534382
This item is not returnable.
View return policy
Description
A suggested positive control is rat spleen tissue lysate. PA5-34382 can be used with blocking peptide PEP-1425.
CLEC7A, also known as dectin-1, is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily and is predominantly expressed on myeloid cells. It is a small glycoprotein type II membrane receptor with an extracellular C-type lectin-like domain fold and a cytoplasmic domain with an immunoreceptor tyrosine-based activation motif (ITAM). CLEC7A functions as a pattern-recognition receptor that recognizes a variety of beta-1,3-linked and beta-1,6-linked glucans from fungi and plants, and in this way plays a role in innate immune response. Upon fungal exposure, CLEC7A activates Syk tyrosine kinase, triggering a massive oxidative burst through the formation of reactive oxygen species.
Specifications
| Dectin 1 | |
| Polyclonal | |
| Unconjugated | |
| Clec7a | |
| Beta-glucan receptor; beta-GR; BGR; CANDF4; CD369; CLEC7A; Clec7b; CLECSF12; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin domain family 7, member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; DECTIN1; dectin-1; Dendritic cell-associated C-type lectin 1; dendritic cell-associated C-type lectin-1; lectin-like receptor 1; RGD1565140; SCARE2; UNQ539/PRO1082 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 502902, 56644, 64581 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6QLQ4, Q9BXN2 | |
| Clec7a | |
| A 17 amino acid synthetic peptide near the center of human CLEC7A. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction