missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Decrosslink Buffer D-Link
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
Description
Compatible Kits include:
- NucleoSpin™ DNA FFPE XS
- NucleoSpin™ 8 / 96 DNA FFPE
- NucleoMag™ DNA FFPE
Specifications
Specifications
| Chemical Name or Material | Decrosslinking buffer |
| Color | Colorless |
| Concentration | 5 to 10% |
| For Use With (Application) | DNA isolation from FFPE samples |
| For Use With (Equipment) | NucleoSpin and NucleoMag DNA FFPE kits |
| pH | 4.5 to 5.0 |
| Physical Form | Liquid |
| Quantity | 30 mL |
| Formulation | Inorganic salts |
| Odor | Odorless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction