missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DECR1 Mouse anti-Human, Clone: 3B2, Novus Biologicals™
Mouse Monoclonal Antibody
£234.00 - £378.00
Specifications
| Antigen | DECR1 |
|---|---|
| Clone | 3B2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1069, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18246901
|
Novus Biologicals
NBP2-59469-50UL |
50 μL |
£234.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18278634
|
Novus Biologicals
NBP2-59469-100UL |
100 μL |
£378.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DECR1 Monoclonal specifically detects DECR1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| DECR1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| 2,4-dienoyl CoA reductase 1, mitochondrial, 4-enoyl-CoA reductase, 4-enoyl-CoA reductase [NADPH], DECR, DECR2,4-dienoyl-CoA reductase [NADPH]2,4-dienoyl-CoA reductase, mitochondrial, EC 1.3.1.34, NADPH, SDR18C1, short chain dehydrogenase/reductase family 18C, member 1 | |
| DECR1 | |
| IgG2b κ | |
| Protein A purified |
| 3B2 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1069, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 1666 | |
| Recombinant human DECR1 (35-335aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title