missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DECR1 Antibody (3B2), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59469PE
This item is not returnable.
View return policy
Description
DECR1 Monoclonal antibody specifically detects DECR1 in Human samples. It is validated for Flow Cytometry
Specifications
| DECR1 | |
| Monoclonal | |
| PE | |
| 2,4-dienoyl CoA reductase 1, mitochondrial, 4-enoyl-CoA reductase, 4-enoyl-CoA reductase [NADPH], DECR, DECR2,4-dienoyl-CoA reductase [NADPH]2,4-dienoyl-CoA reductase, mitochondrial, EC 1.3.1.34, NADPH, SDR18C1, short chain dehydrogenase/reductase family 18C, member 1 | |
| Recombinant human DECR1 (35-335aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 1666 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry | |
| 3B2 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction