missing translation for 'onlineSavingsMsg'
Learn More

Decorin Antibody (DCN/7031R), DyLight 488, Novus Biologicals™

Product Code. 18365072
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365072 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365072 Supplier Bio-Techne Supplier No. NBP314026G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Decorin Monoclonal antibody specifically detects Decorin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Decorin
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DCN/7031R
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B
Host Species Rabbit
Immunogen Recombinant human Decorin protein fragment (around aa212-336) (exact sequence is proprietary) (Uniprot: P07585)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1634
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.