missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Decorin Monoclonal antibody specifically detects Decorin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Decorin |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | DCN/7031R |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B |
| Host Species | Rabbit |
| Immunogen | Recombinant human Decorin protein fragment (around aa212-336) (exact sequence is proprietary) (Uniprot: P07585) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?