missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decorin Antibody (DCN/3523), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79908F
This item is not returnable.
View return policy
Description
Decorin Monoclonal specifically detects Decorin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Decorin | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| DCN | |
| Recombinant human Decorin protein fragment (aa212-336) (exact sequence is proprietary) (Uniprot: P07585) | |
| 0.1 mL | |
| Cancer | |
| Human | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DCN/3523 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction