missing translation for 'onlineSavingsMsg'
Learn More

Decloxizine dihydrochloride, MedChemExpress

Product Code. p-200101790
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16498016

Brand: Medchem Express HYA0075

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Decloxizine dihydrochloride(UCB-1402; NSC289116) is a histamine 1 receptor antagonist.

Decloxizine dihydrochloride(UCB-1402

Specifications

Chemical Name or Material Decloxizine dihydrochloride
Physical Form Solid
CAS 13073-96-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H30Cl2N2O2
For Use With (Application) COVID-19-immunoregulation
Synonym UCB 1402 dihydrochloride
Solubility Information H2O : ≥ 100 mg/mL (241.91 mM) ∣DMSO : ≥ 50 mg/mL (120.95 mM)
SMILES OCCOCCN1CCN(C(C2=CC=CC=C2)C3=CC=CC=C3)CC1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 413.38
Formula Weight 413.38
Percent Purity 98.77%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.