Learn More
Corning™ DeckWorks™ Tip Stations, Sterile
Designed for single and multichannel pipetting stations
£73.54 - £282.70
Specifications
Sterility | Sterile |
---|---|
Color | Natural |
Tip Style | Round |
Packaging | Multi-pack |
For Use With (Equipment) | Universal Fit, Multichannel Pipetting Stations |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11921861
|
Corning™
4144 |
1 to 200 μL |
£73.54
Pack of 960 |
In Stock
Log in to see stock. |
|||||
11911861
|
Corning™
4143 |
0.1 to 10 μL |
£73.54
Pack of 960 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11393665
|
Corning™
4144 |
1 to 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11383665
|
Corning™
4143 |
0.1 to 10 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip stations contain 960 tips, are available in extended-length 10 μL and traditional 200 μL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only. Applications include pre- and post-PCR procedures, cell culture, and assays requiring large tip quantities. Corning DeckWorks tip stations are offered both sterile and nonsterile.
Specifications
Sterile | |
Round | |
Universal Fit, Multichannel Pipetting Stations | |
DeckWorks |
Natural | |
Multi-pack | |
Yes | |
Polypropylene |