Learn More
Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips, Sterile
Designed for quantitative assays in which sample binding can significantly alter results
Brand: Corning™ 4152
Code : 67
Additional Details : Weight : 1.49400kg
Description
Corning DeckWorks low binding tips are available in 10 μL, 200 μL, and 300 μL sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
Specifications
1 to 300 μL | |
Natural | |
Hinged Rack | |
Hinged rack | |
DeckWorks |
Sterile | |
Designed for quantitative assays in which sample binding can significantly alter results | |
Low Retention, Round | |
Yes | |
Polypropylene |