missing translation for 'onlineSavingsMsg'
Learn More

Decernotinib, MedChemExpress

Product Code. 16483306
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483306 25 mg 25mg
16455188 10 mg 10mg
16451407 50 mg 50mg
16450166 2 mg 2mg
16407917 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483306 Supplier Medchem Express Supplier No. HY12469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Decernotinib is a potent, orally active JAK3 inhibitor, with Kis of 2.5, 11, 13 and 11 nM for JAK3, JAK1, JAK2, and TYK2, respectively.

Decernotinib is a potent, orally active JAK3 inhibitor, with Kis of 2.5, 11, 13 and 11 nM for JAK3, JAK1, JAK2, and TYK2, respectively.

Specifications

Chemical Name or Material Decernotinib
Physical Form Solid
CAS 944842-54-0
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C18H19F3N6O
For Use With (Application) COVID-19-immunoregulation
Synonym VX-509 VRT-831509
Solubility Information DMSO : ≥ 50 mg/mL (127.43 mM)
SMILES C[C@](C(NCC(F)(F)F)=O)(CC)NC1=NC(C2=CNC3=NC=CC=C32)=NC=C1
Molecular Weight (g/mol) 392.38
Formula Weight 392.38
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.