missing translation for 'onlineSavingsMsg'
Learn More

Decarboxy Ciprofloxacin, TRC

Product Code. 30135725
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135725 100 mg 100mg
30010424 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135725 Supplier Toronto Research Chemicals Supplier No. TRCD220500100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105394-83-0
Molecular Formula C16 H18 F N3 O
Molecular Weight (g/mol) 287.33
Synonym 1-Cyclopropyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one, Ciprofloxacin Hydrochloride Imp. E (EP), Decarboxylated Ciprofloxacin
IUPAC Name 1-cyclopropyl-6-fluoro-7-piperazin-1-ylquinolin-4-one
SMILES Fc1cc2C(=O)C=CN(C3CC3)c2cc1N4CCNCC4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H18FN3O/c17-13-9-12-14(10-15(13)19-7-4-18-5-8-19)20(11-1-2-11)6-3-16(12)21/h3,6,9-11,18H,1-2,4-5,7-8H2
Formula Weight 287.1434
Chemical Name or Material 1-Cyclopropyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.