missing translation for 'onlineSavingsMsg'
Learn More

Decanoyl chloride, 98%

Product Code. 11484057
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484057 100 g 100g
11494057 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484057 Supplier Thermo Scientific Alfa Aesar Supplier No. A19486.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-13-0 | C10H19ClO | 190.711 g/mol

Decanoyl chloride is used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units. It enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decanoyl chloride is used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units. It enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-13-0
Molecular Formula C10H19ClO
Molecular Weight (g/mol) 190.711
MDL Number MFCD00000771
InChI Key IPIVAXLHTVNRBS-UHFFFAOYSA-N
Synonym capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n
PubChem CID 66982
IUPAC Name decanoyl chloride
SMILES CCCCCCCCCC(=O)Cl

Specifications

Melting Point -35°C
Density 0.925
Boiling Point 232°C
Flash Point 106°C (223°F)
Odor Irritating
Refractive Index 1.441
Quantity 100 g
UN Number UN3265
Beilstein 507055
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 190.71
Percent Purity 98%
Chemical Name or Material Decanoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.