missing translation for 'onlineSavingsMsg'
Learn More

Decanoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog Number 10081880
22.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


21.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
112-13-0
C10H19ClO
190.71
MFCD00000771
IPIVAXLHTVNRBS-UHFFFAOYSA-N
capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n
66982
decanoyl chloride
CCCCCCCCCC(=O)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
112-13-0
C10H19ClO
190.71
MFCD00000771
IPIVAXLHTVNRBS-UHFFFAOYSA-N
capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n
66982
decanoyl chloride
CCCCCCCCCC(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -34.5°C
Color Brown to White
Density 0.9190g/mL
Boiling Point 232.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)8COCl
Refractive Index 1.4400 to 1.4420
Quantity 100 g
Beilstein 02, 356
Specific Gravity 0.919
Solubility Information Solubility in water: 100mg/l (20°C). Other solubilities: hydrolyses in water,450g/L methanol- 500g/L ethanol,500g/L 2-propanol- 500g/L dmf,500g/L acetone- 500g/L dichloromethane
Formula Weight 190.71
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material Decanoyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Decanoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'