missing translation for 'onlineSavingsMsg'
Learn More

Decanoic Acid, TRC

Product Code. 30367137
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367137 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367137 Supplier Toronto Research Chemicals Supplier No. TRCD2125255G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Capric Acid
CAS 334-48-5
Molecular Formula C10 H20 O2
Quantity 5 g
Synonym 1-Decanoic acid, 1-Nonanecarboxylic acid, Caprates, Capric acid, Caprinic acid, Caprynic acid, Decoic acid, Decylic acid, Edenor C 10-98-100, Emery 659, Lunac 10-95, Lunac 10-98, Lunac 10-98E, NAA 102, NSC 5025, Palamac 99-10, Palmac 99-10, Prifac 2906, Prifac 296, n-Capric acid, n-Decanoic acid, n-Decoic acid, n-Decylic acid
InChI Formula InChI=1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)
SMILES CCCCCCCCCC(=O)O
IUPAC Name decanoic acid
Molecular Weight (g/mol) 172.26
Formula Weight 172.1463
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.