missing translation for 'onlineSavingsMsg'
Learn More

Decanoic acid, MedChemExpress

Product Code. 16443449
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443449 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16443449 Supplier Medchem Express Supplier No. HYW015309
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Decanoic acid, a component of medium chain triclycerides, is a brain-penetrant and non-competitive inhibitor of AMPA receptor. Decanoic acid has antiseizure effects.

Decanoic acid, a component of medium chain triclycerides, is a brain-penetrant and non-competitive inhibitor of AMPA receptor. Decanoic acid has antiseizure effects.

Specifications

Chemical Name or Material Decanoic acid
CAS 334-48-5
Color Colorless
Purity Grade Notes Research
Molecular Formula C10H20O2
Quantity 1 g
Solubility Information DMSO : ≥ 130 mg/mL (754.63 mM)
SMILES CCCCCCCCCC(O)=O
Molecular Weight (g/mol) 172.27
Formula Weight 172.27
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.