missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decanoic-10,10,10-d3 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Decanoic-10,10,10-d3 Acid |
| CAS | 102611-15-4 |
| Molecular Formula | C10 D3 H17 O2 |
| Quantity | 0.25 g |
| Synonym | 1-Decanoic Acid-d3, 1-Nonanecarboxylic Acid-d3, Capric Acid-d3, Caprinic Acid-d3, Caprynic Acid-d3, Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI), Decanoic-10,10,10-d3 acid (9 CI) |
| InChI Formula | InChI=1 S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCCCCCCC(=O)O |
| IUPAC Name | 10,10,10-trideuteriodecanoic acid |
| Molecular Weight (g/mol) | 175.28 |
| Formula Weight | 175.1652 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction