Product Code. 30136389
Click to view available options
Quantity:
25 mL
100 mL
250 mL
Unit Size:
100mL
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
30136389
250 mL
250mL
30133135
100 mL
100mL
17913425
25 mL
25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
30136389
Supplier
Toronto Research Chemicals
Supplier No.
TRCD212510250ML
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Chemical Identifiers
CAS
124-18-5
Molecular Formula
C10 H22
Molecular Weight (g/mol)
142.28
Synonym
Decane (8CI, 9CI, ACI), Cactus Normal Paraffin N 10, NSC 8781, n-Decane
IUPAC Name
decane
SMILES
CCCCCCCCCC
Specifications
Quantity
250 mL
InChI Formula
InChI=1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3
Formula Weight
142.1722
Chemical Name or Material
n-Decane
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?