missing translation for 'onlineSavingsMsg'
Learn More

Decane, TRC

Product Code. 30133135
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
Unit Size:
100mL
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133135 100 mL 100mL
30136389 250 mL 250mL
17913425 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 30133135 Supplier Toronto Research Chemicals Supplier No. TRCD212510100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10 H22
Molecular Weight (g/mol) 142.28
Synonym Decane (8CI, 9CI, ACI), Cactus Normal Paraffin N 10, NSC 8781, n-Decane
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Quantity 100 mL
InChI Formula InChI=1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3
Formula Weight 142.1722
Chemical Name or Material n-Decane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.