missing translation for 'onlineSavingsMsg'
Learn More

Decanal, TRC

Product Code. 30136742
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136742 50 g 50g
30008698 100 g 100g
17905085 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136742 Supplier Toronto Research Chemicals Supplier No. TRCD21250550G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-31-2
Molecular Formula C10 H20 O
Molecular Weight (g/mol) 156.27
Synonym Decanal, Capraldehyde, Capric aldehyde, Caprinaldehyde, Caprinic aldehyde, Decaldehyde, Decanaldehyde, Decyl aldehyde, Decylic aldehyde, NSC 6087, n-Decaldehyde, n-Decanal, n-Decyl aldehyde
IUPAC Name decanal
SMILES CCCCCCCCCC=O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h10H,2-9H2,1H3
Formula Weight 156.1514
Chemical Name or Material Decanal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.