missing translation for 'onlineSavingsMsg'
Learn More

Decanal, 96%

Product Code. 11474773
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474773 500 mL 500mL
11464773 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474773 Supplier Thermo Scientific Alfa Aesar Supplier No. A11656.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-31-2 | C10H20O | 156.269 g/mol

Decanal is used in fragrances and flavoring and it is an important component in citrus along with octanal, citral, and sinensal. It is also used in Air Care Products, Laundry and Dishwashing Products, Personal Care Products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decanal is used in fragrances and flavoring and it is an important component in citrus along with octanal, citral, and sinensal. It is also used in Air Care Products, Laundry and Dishwashing Products, Personal Care Products.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-31-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00007031
InChI Key KSMVZQYAVGTKIV-UHFFFAOYSA-N
Synonym decyl aldehyde, capraldehyde, caprinaldehyde, decanaldehyde, n-decyl aldehyde, capric aldehyde, 1-decanal, n-decanal, n-decaldehyde, 1-decyl aldehyde
PubChem CID 8175
ChEBI CHEBI:31457
IUPAC Name decanal
SMILES CCCCCCCCCC=O

Specifications

Melting Point -5°C
Density 0.828
Boiling Point 207°C to 209°C
Flash Point 85°C (185°F)
Assay Percent Range 96%
Linear Formula CH3(CH2)8CHO
Refractive Index 1.428
Quantity 500 mL
Beilstein 1362530
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 156.27
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Decanal
Show More Show Less
compliance-icons
Product Identifier
  • Decanal
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Decanal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.