missing translation for 'onlineSavingsMsg'
Learn More

Decamethylferrocene, 99%, Thermo Scientific Chemicals

Product Code. 11398188
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398188 1 g 1g
11308198 250 mg 250mg
11388188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11398188 Supplier Thermo Scientific Alfa Aesar Supplier No. 039406.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Decamethylferrocene is used as a superior internal reference redox standard to ferrocene in voltammetric and amperometric titrations. It is used as photocathode for the detection of barium fluoride scintillation light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12126-50-0
Molecular Formula C20H30Fe
Molecular Weight (g/mol) 326.31
MDL Number MFCD00058736
InChI Key VKRKSGODPDKGAD-UHFFFAOYSA-N
Synonym Bis(pentamethylcyclopentadienyl)iron(II)
SMILES Cc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Fe]2345

Specifications

Melting Point 298°C to 300°C (sublimation)
Odor Odorless
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 326.31
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Decamethylferrocene
compliance-icons
Product Identifier
  • Decamethylferrocene
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.