missing translation for 'onlineSavingsMsg'
Learn More

Decamethylferrocene, 99%, Thermo Scientific Chemicals

Product Code. p-7050757
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11308198

Brand: Thermo Scientific Alfa Aesar 039406.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Decamethylferrocene is used as a superior internal reference redox standard to ferrocene in voltammetric and amperometric titrations. It is used as photocathode for the detection of barium fluoride scintillation light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12126-50-0
Molecular Formula C20H30Fe
Molecular Weight (g/mol) 326.31
MDL Number MFCD00058736
InChI Key VKRKSGODPDKGAD-UHFFFAOYSA-N
Synonym Bis(pentamethylcyclopentadienyl)iron(II)
SMILES Cc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Fe]2345

Specifications

Melting Point 298°C to 300°C (sublimation)
Odor Odorless
Quantity 250 mg
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 326.31
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Decamethylferrocene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.