missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decamethylferrocene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Decamethylferrocene is used as a superior internal reference redox standard to ferrocene in voltammetric and amperometric titrations. It is used as photocathode for the detection of barium fluoride scintillation light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 298°C to 300°C (sublimation) |
Odor | Odorless |
Quantity | 250 mg |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 326.31 |
Percent Purity | 99% |
Physical Form | Crystalline |
Chemical Name or Material | Decamethylferrocene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'