missing translation for 'onlineSavingsMsg'
Learn More

Decamethylcyclopentasiloxane, TRC

Product Code. 30136083
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136083 50 g 50g
30132789 100 g 100g
17957814 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136083 Supplier Toronto Research Chemicals Supplier No. TRCD21244050G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-02-6
Molecular Formula C10 H30 O5 Si5
Molecular Weight (g/mol) 370.77
Synonym Cyclopentasiloxane, 2,2,4,4,6,6,8,8,10,10-decamethyl-, Cyclopentasiloxane, decamethyl- (6CI,8CI,9CI), 2,2,4,4,6,6,8,8,10,10-Decamethylcyclopentasiloxane, Botanisil CP 33, Cyclic dimethylsiloxane pentamer, Cyclo-decamethylpentasiloxane, Cyclomethicone pentamer 245, Cyclopentadimethylsiloxane, Cyclopentasiloxane, D 5 (siloxane), D5, DC 2-5252C, DC 245, DC 345, DC 345 Fluid, Decamethylcyclopentasiloxane, Decamethylcylopentasiloxane, Decamethylpentacyclosiloxane, Dimethylsiloxane pentamer, Dow Corning 2-5252C, Dow Corning 245, Dow Corning 245 Fluid, Dow Corning 345, Dow Corning 345 Fluid, Dow Corning 345EU, Execol D 5, KF 7312T, KF 955, KF 995, LS 9000, Mirasil CM 5, NUC Silicone VS 7158, Pentacyclomethicone, SF 1202, SH 245, SH 245 (siloxane), SH 245 Fluid, Silbione 70045V5, Silbione V 5, Silicon Plus α, Silicone SF 1202, Siloxane D 5, TFS 405, TSF 405, TSF 405A, TSF 465, Tego Polish Additive 5, Union Carbide 7158 Silicone Fluid, VS 7158, Volasil 245, Volatile Silicone Fluid 345, Xiameter PMX, Xiameter PMX 0245
IUPAC Name 2,2,4,4,6,6,8,8,10,10-decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
SMILES C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H30O5Si5/c1-16(2)11-17(3,4)13-19(7,8)15-20(9,10)14-18(5,6)12-16/h1-10H3
Formula Weight 370.094
Chemical Name or Material Cyclomethicone 5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.