missing translation for 'onlineSavingsMsg'
Learn More

Decamethylcyclopentasiloxane, SPEX CertiPrep™

Codice prodotto. 15246126 Sfoglia Tutto SPEX Certiprep Prodotti
Cambia vista
Click to view available options
Quantity:
1 mL
Dimensione della confezione:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
15246126 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativo: Questo prodotto contiene una sostanza elencata nell'allegato XIV del regolamento REACH (CE) n. 1907/2006. Tutti i clienti devono compilare una dichiarazione prima della consegna.
Codice prodotto. 15246126 Fornitore SPEX Certiprep N. del fornitore S1110

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativo: Questo prodotto contiene una sostanza elencata nell'allegato XIV del regolamento REACH (CE) n. 1907/2006. Tutti i clienti devono compilare una dichiarazione prima della consegna.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Decamethylcyclopentasiloxane541-02-61000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifica

Packaging 2mL amber prescored glass ampule
CAS 541-02-6, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C10H30O5Si5
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key XMSXQFUHVRWGNA-UHFFFAOYSA-N
SMILES C[Si]1(O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)C
IUPAC Name 2,2,4,4,6,6,8,8,10,10-decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
Solvent or Matrix Methanol-P&T
Molecular Weight (g/mol) 370.77
PubChem CID 10913
Quantity 1 mL
Chemical Name or Material Decamethylcyclopentasiloxane
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons compliance-icons
Identificativo prodotto
  • Decamethylcyclopentasiloxane
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Indicazioni di pericolo
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informazioni supplementari
  • MIXTURE LIST-Contain: methanol

Product intended for laboratory use only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.