missing translation for 'onlineSavingsMsg'
Learn More

DecahydronapHtalene, 99+%, Pure, Fisher Chemical

Product Code. 11408163 Shop All Fisher Chemical Products
Change view
Click to view available options
2.5L, Amber glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11408163 2.5L, Amber glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408163 Supplier Fisher Chemical Supplier No. D/0052/17
Quantity Price
16 £139.36 / 2.5 L
1 £163.95 / 2.5 L
4 £147.56 / 2.5 L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 91-17-8
Melting Point -31°C
Color Colorless
pH 7
Boiling Point 187°C
Packaging Amber glass bottle
Molecular Formula C10H18
MDL Number 4130
Quantity 2.5 L
Vapor Density 4.8
Formula Weight 138.25g/mol
Vapor Pressure 1.3mbar 22
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Decahydronaphthalene, mixture of cis and trans
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.