missing translation for 'onlineSavingsMsg'
Learn More

Decafluorobenzophenone, 97%, Thermo Scientific™

Product Code. 15590514 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15590514 5 g 5g
15580514 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15590514 Supplier Thermo Scientific Acros Supplier No. 146590050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 853-39-4
Molecular Formula C13F10O
Molecular Weight (g/mol) 362.13
InChI Key WWQLXRAKBJVNCC-UHFFFAOYSA-N
Synonym decafluorobenzophenone, benzophenone, decafluoro, perfluorobenzophenone, methanone, bis pentafluorophenyl, bis perfluorophenyl methanone, bis pentafluorophenyl methanone, bis 2,3,4,5,6-pentafluorophenyl methanone, di2,3,4,5,6-pentafluorophenyl ketone, acmc-209q5n
PubChem CID 70068
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)methanone
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)C2=C(C(=C(C(=C2F)F)F)F)F

Specifications

Melting Point 91.0°C to 95.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6F5)2CO
Quantity 5 g
Formula Weight 362.13
Percent Purity 97%
Chemical Name or Material Decafluorobenzophenone
compliance-icons
Product Identifier
  • Decafluorobenzophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.