missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DECACELL™ Cuvettes

Product Code. 17886905 Shop All Thermo Scientific Products
Product Code. 17886905 Supplier Thermo Scientific™ Supplier No. 986540TS

Please to purchase this item. Need a web account? Register with us today!

Reduce reagent consumption and cost per analysis and improve test precision, accuracy, and reliability.

Improve test precision, accuracy, reliability, and cost per analysis with the low-cost, disposable DECACELL™ cuvettes.

Cuvettes for faster, more cost-effective analyses
  • Reduce sample and reagent consumption with low-volume, 2—240 μL cuvettes
  • Improve throughput by minimizing the need for cleaning
  • Minimize carryover and cross contamination
  • Reduce waste generation
  • Achieve lower cost per analysis

Specifications

Type Cuvette
Description DECACELL™ Cuvettes
Color Clear
Disposable Single-use
Capacity (Metric) 2 to 240 μL
Quantity One box of 540 cuvettes with 10 cells each
For Use With (Equipment) Thermo Scientific™ Gallery™ and Gallery™ Plus Discrete Analyzers
For Use With (Application) Automated Wet Chemical Analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.