missing translation for 'onlineSavingsMsg'
Learn More

Decacarbonyldirhenium, 96%, Thermo Scientific Chemicals

Product Code. 11394769
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394769 250 mg 250mg
11384769 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394769 Supplier Thermo Scientific Alfa Aesar Supplier No. 062108.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14285-68-8 | Molecular Formula: C10O10Re2 | Molecular Weight: 652.51 g/mol

Decacarbonyldirhenium is a key starting material used for the synthesis of other rhenium carbonyl complexes. It plays an important role in metathesis, reforming and hydrogenation reactions. It is used as a catalyst to promote silation of alcohols and in the preparion of silyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14285-68-8
Molecular Formula C10O10Re2
Molecular Weight (g/mol) 652.51
MDL Number MFCD00011198
InChI Key ZIZHEHXAMPQGEK-UHFFFAOYSA-N
Synonym carbon monoxide; rhenium 2+, decacarbonyldirhenium 250mg
PubChem CID 498777
SMILES [O]#C[Re](C#[O])(C#[O])(C#[O])(C#[O])[Re](C#[O])(C#[O])(C#[O])(C#[O])C#[O]

Specifications

Melting Point 170°C (decomposition)
Color White
Density 2.87g/mL
Linear Formula Re2(CO)10
Quantity 250 mg
UN Number UN3466
IUPAC Name {[1,1,1,1,2,2,2,2,2-nonakis(hydroxymethyl)dirhenium-1-yl]methylidyne}-λ³-oxidane
Formula Weight 652.52
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Decacarbonyldirhenium
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.