missing translation for 'onlineSavingsMsg'
Learn More

DEC2/SHARP1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£151.00 - £359.00

Specifications

Antigen DEC2/SHARP1
Dilution Western Blot 1:1000-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18613530
View Documents
Novus Biologicals
NBP2-92031-0.02ml
0.02 mL
£151.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18699230
View Documents
Novus Biologicals
NBP2-92031-0.1ml
0.1 mL
£359.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DEC2/SHARP1 Polyclonal antibody specifically detects DEC2/SHARP1 in Human, Rat samples. It is validated for Western Blot
Specifications

Specifications

DEC2/SHARP1
Western Blot
Unconjugated
Rabbit
Cancer, Endocrinology, Neuroscience, Signal Transduction
PBS with 50% glycerol, pH7.3.
79365
IgG
Affinity purified
Western Blot 1:1000-1:2000
Polyclonal
Purified
RUO
Human, Rat
basic helix-loop-helix domain containing, class B, 3, basic helix-loop-helix family, member e41, bHLHb3, BHLHB3class E basic helix-loop-helix protein 41, bHLHe41, Class B basic helix-loop-helix protein 3, DEC2Enhancer-of-split and hairy-related protein 1, Differentially expressed in chondrocytes protein 2, SHARP-1, SHARP1hDEC2
Recombinant fusion protein containing a sequence corresponding to amino acids 120-280 of human BHLHE41 (NP_110389.1). LKSPIQSDLDAFHSGFQTCAKEVLQYLSRFESWTPREPRCVQLINHLHAVATQFLPTPQLLTQQVPLSKGTGAPSAAGSAAAPCLERAGQKLEPLAYCVPVIQRTQPSAELAAENDTDTDSGYGGEAEARPDREKGKGAGASRVTIKQEPPGEDSPAPKRM
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.