missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25021-1-AP-150UL
This item is not returnable.
View return policy
Description
Dec1, which is also known as Stra13 or Sharp2, is a basic helix-loop-helix (bHLH) transcription factorthat has beenfound to play an important role in circadian system. Dec1contains one basic helix-loop-helix (bHLH) domain, one Orange domain and is reported to be expressed in Treg cells. It is also reported that Dec1 is required for long-term maintece of Treg cells after adoptive transfer to suppress effector T cell-mediated inflammation. Dec1 positively regulates CD25 expression in association with Runx 1.Specifications
| DEC1 | |
| Polyclonal | |
| Unconjugated | |
| DEC1 | |
| CTS9, DEC1, deleted in esophageal cancer 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 50514 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9P2X7 | |
| DEC1 | |
| DEC1 Fusion Protein Ag20192 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction