missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEC-205/CD205 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | Renal Cell Carcinoma (gp200) |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247141
|
Novus Biologicals
NBP2-58339 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654147
|
Novus Biologicals
NBP2-58339-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DEC-205/CD205 Polyclonal specifically detects DEC-205/CD205 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Renal Cell Carcinoma (gp200) | |
| Polyclonal | |
| Rabbit | |
| Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers, Vision | |
| CD205LY-75, CLEC13BLy-75, C-type lectin domain family 13 member B, DEC-205CD205 antigen, GP200-MR6, lymphocyte antigen 75 | |
| LY75 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 4065 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QRHRLHLAGFSSVRYAQGVNEDEIMLPSFH | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title