Learn More
Novus Biologicals™ DEC-205/CD205 Antibody (CDX-1401) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28277-50ug
 View more versions of this product
Product Code. 30047870
  • £162.00 / 50µg
Description
DEC-205/CD205 Monoclonal antibody specifically detects DEC-205/CD205 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DEC-205/CD205
Monoclonal
Unconjugated
O60449
Human
Protein A purified
RUO
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
CDX-1401
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD205;DCL-1 fusion;DEC-205;gp200;Ly-75;Lymphocyte antigen 75;membrane antigen MN;RP23-10L17.4
LY75/ CD205/DEC-205
50 μg
Primary
Human
Purified