missing translation for 'onlineSavingsMsg'
Learn More

Deblock trichloroacetic acid soln., 3% w/w in dichloromethane

Product Code. 15440267
Click to view available options
Quantity:
450 mL
2.5 L
Unit Size:
2.5 L
450mL
This item is not returnable. View return policy

Product Code. 15440267

Brand: Thermo Scientific Alfa Aesar J62821.AO

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deblock trichloroacetic acid soln.
Color Colorless
MDL Number MFCD00004177
UN Number UN2810
Concentration 3% w/w in dichloromethane
Density 1.325
Synonym DEBLK soln.
Quantity 450 mL
Physical Form Liquid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.