missing translation for 'onlineSavingsMsg'
Learn More

Death Ligand Signal Enhancer Antibody, DyLight 650, Novus Biologicals™

Product Code. 18405597 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405597 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405597 Supplier Novus Biologicals Supplier No. NBP281912C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Death Ligand Signal Enhancer Polyclonal antibody specifically detects Death Ligand Signal Enhancer in Human, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Death Ligand Signal Enhancer
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias DAP3 Binding Cell Death Enhancer 1, DAP3-Binding Cell Death Enhancer 1, DAP3-Binding Cell Death Enhancer 1, Long Form, Death Ligand Signal Enhancer, DELE, DELE1, DELE1(L), hypothetical protein LOC9812, KIAA0141
Host Species Rabbit
Immunogen Death Ligand Signal Enhancer antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human Death Ligand Signal Enhancer. The immunogen is located within the first 50 amino acids of Death Ligand Signal Enhancer. Amino Acid Squence: RVTPKSTSPDGPQTTSST
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9812
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.