missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEAF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530388
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1. Predicted reactivity: Mouse (98%), Rat (98%), Chicken (89%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HNF1A is a transcription factor required for the expression of several liver-specific genes. This protein functions as a homodimer and binds to the inverted palindrome 5'-GTTAATNATTAAC-3'.
Specifications
| DEAF1 | |
| Polyclonal | |
| Unconjugated | |
| Deaf1 | |
| AU042387; C230009B13Rik; DEAF1; DEAF-1 related transcriptional regulator (NUDR); DEAF1 transcription factor; DEAF1, transcription factor; deformed epidermal autoregulatory factor 1 (Drosophila); deformed epidermal autoregulatory factor 1 homolog; MRD24; Nuclear DEAF-1-related transcriptional regulator; NUDR; SPN; suppressin; Zinc finger MYND domain-containing protein 5; ZMYND5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10522 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O75398 | |
| Deaf1 | |
| Recombinant protein encompassing a sequence within the center region of human DEAF1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction