missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEAF1 Monoclonal Antibody (3E6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521069
This item is not returnable.
View return policy
Description
Peptide Sequence: TALQIGDSLN TEKATLIVVH TDGSIVETTG LKGPAAPLTP GPQSPPTPLA PGQEKGGTKY NWDPSVYDSE LPVRCRNISG TLYKNRLGSG.
HNF1A is a transcription factor required for the expression of several liver-specific genes. This protein functions as a homodimer and binds to the inverted palindrome 5'-GTTAATNATTAAC-3'.
Specifications
| DEAF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O75398 | |
| Deaf1 | |
| DEAF1 (NP_066288.2, 133 a.a. ~ 222 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 3E6 | |
| Unconjugated | |
| Deaf1 | |
| AU042387; C230009B13Rik; DEAF1; DEAF-1 related transcriptional regulator (NUDR); DEAF1 transcription factor; DEAF1, transcription factor; deformed epidermal autoregulatory factor 1 (Drosophila); deformed epidermal autoregulatory factor 1 homolog; MRD24; Nuclear DEAF-1-related transcriptional regulator; NUDR; SPN; suppressin; Zinc finger MYND domain-containing protein 5; ZMYND5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10522 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction