missing translation for 'onlineSavingsMsg'
Learn More

DEAE-Cellulose, (approx. 70% moisture content), TRC

Product Code. 30003598
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003598 1 g 1g
30003488 50 g 50g
30008527 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003598 Supplier Toronto Research Chemicals Supplier No. TRCD1990501G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9013-34-7
Molecular Formula C18H38N2O6
Molecular Weight (g/mol) 378.5
Synonym Cellulose-DEAE, Diethylaminoethyl-cellulose
IUPAC Name (2R,3R,4S,5S,6R)-3-[2-(diethylamino)ethoxy]-6-[2-(diethylamino)ethoxymethyl]oxane-2,4,5-triol
SMILES CCN(CC)CCOC[C@@H]1[C@H]([C@@H]([C@H]([C@@H](O1)O)OCCN(CC)CC)O)O

Tekniske data

Quantity 1 g
InChI Formula InChI=1S/C18H38N2O6/c1-5-19(6-2)9-11-24-13-14-15(21)16(22)17(18(23)26-14)25-12-10-20(7-3)8-4/h14-18,21-23H,5-13H2,1-4H3/t14-,15-,16+,17-,18-/m1/s1
Chemical Name or Material DEAE-Cellulose
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.