missing translation for 'onlineSavingsMsg'
Learn More

Deacetylcephalothin Lactone, TRC

Product Code. 30007846
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007846 10 mg 10mg
30008231 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007846 Supplier Toronto Research Chemicals Supplier No. TRCD19821510MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10590-10-0
Synonym N-[(5aR,6R)-1,4,5a,6-Tetrahydro-1,7-dioxo-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazin-6-yl]-2-thiopheneacetamide∣2-Thiopheneacetamide, N-(1,4,5a,6-tetrahydro-1,7-dioxo-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazin-6-yl)-, (5aR-trans)- (8CI)∣5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-(hydroxymethyl)-8-oxo-7-[2-(2-thienyl)acetamido]-, γ-lactone (7CI)∣3H,7H-Azet∣(5aR,6R)-6-[(Thiophen-2-ylacetyl)amino]-5a,6-dihydro-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazine-1,7(4H)-dioneo[2,1-b]furo[3,4-d][1,3]thiazine, 2-thiopheneacetamide deriv.∣Deacetylcephalothin lactone∣(5aR,6R)-6-[[(thiophen-2-yl)acetyl]amino]-5a,6-dihydro-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazine-1,7(4H)-dione∣Cefalotin lactone∣Cefoxitin Sodium Imp. C (EP)∣Cefalotin EP Impurity D∣3-(hydroxymethyl)-8-oxo-7-[2-(2-thienyl)acetamido]-5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid γ-Lactone∣Deacetylcephalothin Lactone∣3H,7H-Azeto[2,1-b]furo[3,4-d][1,3]thiazine, 2-thiopheneacetamide Deriv.∣(5aR-trans)-N-(1,4,5a,6-tetrahydro-1,7-dioxo-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazin-6-yl)-2-Thiopheneacetamide

Specifications

Format Neat
Molecular Weight (g/mol) 336.39
Product Type Impurity
Quantity 10 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.