missing translation for 'onlineSavingsMsg'
Learn More

De(methypiperazinyl) Sildenafil Dimer Impurity, TRC

Product Code. 30357436
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357436 25 mg 25mg
30355040 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357436 Supplier Toronto Research Chemicals Supplier No. TRCD23121525MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346603-48-2
Molecular Formula C34H38N8O6S
Molecular Weight (g/mol) 686.78
Synonym 5-[2-Ethoxy-5-[4-[4-ethoxy-3-(1-methyl-7-oxo-3-propyl-4,7-dihydro-1H-pyrazolo[4,3-d]pyrimidin-5-yl)phenyl]sulfonyl]phenyl]-1-methyl-3-n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCC1=NN(C2=C1NC(=NC2=O)C3=C(C=CC(=C3)S(=O)(=O)C4=CC(=C(C=C4)OCC)C5=NC(=O)C6=C(N5)C(=NN6C)CCC)OCC)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C34H38N8O6S/c1-7-11-23-27-29(41(5)39-23)33(43)37-31(35-27)21-17-19(13-15-25(21)47-9-3)49(45,46)20-14-16-26(48-10-4)22(18-20)32-36-28-24(12-8-2)40-42(6)30(28)34(44)38-32/h13-18H,7-12H2,1-6H3,(H,35,37,43)(H,36,38,44)
Chemical Name or Material De(methypiperazinyl) Sildenafil Dimer Impurity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.