missing translation for 'onlineSavingsMsg'
Learn More

De(carboxymethoxy) Cetirizine Acetic Acid Dihydrochloride, TRC

Product Code. 30138430
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138430 500 mg 500mg
17984855 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138430 Supplier Toronto Research Chemicals Supplier No. TRCD215505500MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000690-91-4
IUPAC Name 2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]acetic acid;dihydrochloride

Specifications

Molecular Formula C19 H21 Cl N2 O2 . 2 Cl H
Quantity 500 mg
Synonym (RS)-2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]acetic Acid Dihydrochloride, Cetirizine Dihydrochloride Imp. B (EP) as Dihydrochloride
InChI Formula InChI=1S/C19H21ClN2O2.2ClH/c20-17-8-6-16(7-9-17)19(15-4-2-1-3-5-15)22-12-10-21(11-13-22)14-18(23)24;;/h1-9,19H,10-14H2,(H,23,24);2*1H
SMILES Cl.Cl.OC(=O)CN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
Molecular Weight (g/mol) 417.76
Formula Weight 416.0825
Percent Purity >95%
Chemical Name or Material (RS)-2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]acetic Acid Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.