missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX6 Recombinant Rabbit Monoclonal Antibody (6F9I6)

DDX6 Recombinant Rabbit Monoclonal Antibody (ARC1673)
Brand: Invitrogen™ MA535825
This item is not returnable.
View return policy
Description
DDX6 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a member of the DEAD box protein family. The protein is an RNA helicase found in P-bodies and stress granules, and functions in translation suppression and mRNA degradation. It is required for microRNA-induced gene silencing.
Specifications
| DDX6 | |
| Recombinant Monoclonal | |
| 0.53 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26196, P54823 | |
| DDX6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 101-200 of human DDX6 (P26196). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 6F9I6 | |
| Unconjugated | |
| DDX6 | |
| 1110001P04Rik; ATP-dependent RNA helicase p54; DDX6; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 6; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 6; DEAD (Asp-Glu-Ala-Asp) box helicase 6; DEAD (Asp-Glu-Ala-Asp) box polypeptide 6; DEAD box protein 6; DEAD box-6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD); DEAD-box helicase 6; DEAD-box protein 6; E230023J21Rik; HLR2; mRCK/P54; Oncogene RCK; oncogene RCK homolog; p54; probable ATP-dependent RNA helicase DDX6; RCK; RGD1564560 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13209, 1656, 500988 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction